Learn More
CD25 Mouse anti-Human, APC-Cy7, Clone: M-A251, BD
Mouse Monoclonal Antibody
$182.80 - $372.00
Specifications
Antigen | CD25 |
---|---|
Clone | M-A251 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC-Cyanine7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB561782
|
BD Biosciences
561782 |
25 Tests |
Each for $182.80
|
|
|||||
BDB557753
|
BD Biosciences
557753 |
100 Tests |
Each for $372.00
|
|
|||||
Description
The M-A251 monoclonal antibody specifically binds to the 55 kDa type I transmembrane glycoprotein known as the low-affinity interleukin-2 receptor alpha chain subunit (IL-2Rα). CD25 is expressed on regulatory T cells and on activated lymphocytes (T and B) and monocytes. It associates with the IL-2Rβ/CD122 and the IL-2Rγ/CD132 receptor chains to form the high-affinity IL-2R complex. CD25 expression on T and B lymphocytes is upregulated by antigenic or mitogenic stimulation. Soluble CD25/IL-2Rα is produced as a consequence of lymphocyte stimulation and is found in biological fluids following inflammatory responses.
Specifications
CD25 | |
Flow Cytometry | |
APC-Cyanine7 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
M-A251 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.