Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD25 Mouse anti-Human , BUV615, Clone: 2A3, BD Biosciences™

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 612996

 View more versions of this product

Catalog No. BDB612996


Only null left
Add to Cart

Description

Description

CD25 Monoclonal antibody specifically detects CD25 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD25
Monoclonal
Brilliant Ultraviolet 615
IL-2R; IL2RA; IL-2Rα; TCGFR; TAC antigen; p55
Human Phytohemagglutinin-activated T Cells
RUO
Human
IgG1 κ
Flow Cytometry
2A3
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.