Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD25 Rat anti-Mouse, R-PE, Clone: 7D4, Southern Biotech™

Rat Monoclonal Antibody
Supplier: Southern Biotechnology Associates 159509L
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD25 | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
IL-2 dependent BALB/c helper T cell clone HT2 | |
RUO | |
Mouse | |
IgM |
Flow Cytometry | |
7D4 | |
R-PE | |
Rat | |
0.2 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction