Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD253 Mouse anti-Human, Brilliant Violet 786, Clone: RIK-2, BD Optibuild™

Catalog No. BDB743723 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743723 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743723 Supplier BD Biosciences Supplier No. 743723
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The RIK-2 monoclonal antibody specifically binds to CD253, also known as TRAIL (TNF-Related Apoptosis-Inducing Ligand) and Apo2 ligand (APO-2L). CD253 is a member of the Tumor Necrosis Factor Superfamily and is encoded by the TNFSF10 gene. CD253 is a type II membrane protein that may be expressed as a soluble as well as full-length, cell surface-associated protein. Both surface and soluble forms of TRAIL rapidly induce apoptosis in a wide range of tumor cell lines but not normal tissue. TRAIL is expressed by activated T cells, NK cells, dendritic cells, monocytes and a variety of non-lymphoid cells. TRAIL can bind to and exert apoptosis through DR4 (TRAIL-R1) and DR5 (TRAIL-R2) receptors. It can also bind to decoy receptors, including DcR1/TRID/TRAIL-R3 and DcR2/TRUNDD/TRAIL-R4, and possibly OPG/TNFRSF11B, which may serve to regulate TRAIL activity. TRAIL has been shown to be involved in T cell-mediated cytotoxicity, but its mechanism of action remains to be fully elucidated. The RIK-2 clone was selected based on its ability to block TRAIL-mediated cytotoxic activity.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD253
Classification Monoclonal
Clone RIK-2
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TNFSF10, TRAIL, APO-2L, TL2
Host Species Mouse
Immunogen Human TRAIL Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.