Learn More
CD257 (BAFF, BLyS) Monoclonal Antibody (1D6), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$198.00 - $419.00
Specifications
Antigen | CD257 (BAFF, BLyS) |
---|---|
Clone | 1D6 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
17-901-741
![]() |
Invitrogen™
17901741 |
25 Tests |
Each for $198.00
|
|
|||||
17-901-742
![]() |
Invitrogen™
17901742 |
100 Tests |
Each for $419.00
|
|
|||||
Description
The 1D6 MAb reacts with human BLyS (B lymphocyte stimulator), also known as BAFF (B cell activating factor from the TNF family), TALL-1 (TNF and apoptosis leukocyte-expressed ligand-1), THANK (TNF homologue that activates apoptosis, nuclear factor KappaB and c-Jun NH2-terminal kinase) and zTNF4. BLyS, a member of the TNF family, is a type II membrane protein that exists in both membrane-bound and soluble forms. Expression of membrane-bound BLyS on monocytes lineage is regulated by IFN gamma. BLyS acts as a potent B cell growth factor and costimulator of Ig production. BLyS transgenic mice develop severe B cell hyperplasia and autoimmune lupus-like disease characterized by the presence of autoantibodies against nuclear antigens and immune complexes deposited in the kidney. Data from animal models imply a role for BLyS in human autoimmune diseases. To date BCMA, TACI and BR3 have been identified as receptors for BLyS. APRIL, another TNF family member, and BLyS can form active heterotrimeric molecules when coexpressed and these circulating heterotrimers are present in serum samples from patients with systemic immune-based rheumatic diseases. Furthermore, it has been reported that BLyS and APRIL, share BCMA and TACI as their receptors.
BAFF is a membrane bound cytokine expressed by B cell lineage cells. The soluble form of the cytokine binds to the receptors TNFRSF13B/TACI, TNFRSF17/BCMA, and TNFRSF13C/BAFFR. BAFF is a potent B cell activator, and important for the proliferation and differentiation of B cells. BAFF is also expressed by T lymphocytes and has been shown to promote T cell activation and survival. Isoforms representing alternatively spliced, and glycosylated variants have been identified.Specifications
CD257 (BAFF, BLyS) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
ApoL related ligand TALL-1; b cell-activating factor; B lymphocyte stimulator; BAFF; B-cell-activating factor; B-lymphocyte stimulator; BLYS; CD257; D8Ertd387e; delta BAFF; Delta4 BAFF; dendritic cell-derived TNF-like molecule; DTL; RGD1561519; TALL1; TALL-1; THANK; TNF- and APOL-related leukocyte expressed ligand 1; TNF and ApoL-related leukocyte expressed ligand 1; TNF homolog that activates apoptosis; Tnfsf13b; TNFSF20; TNLG7A; tumor necrosis factor (ligand) superfamily member 13b; tumor necrosis factor (ligand) superfamily, member 13b; tumor necrosis factor (ligand) superfamily, member 20; tumor necrosis factor ligand 7A; tumor necrosis factor ligand superfamily member 13B; Tumor necrosis factor ligand superfamily member 13b, membrane form; Tumor necrosis factor ligand superfamily member 13b, soluble form; tumor necrosis factor superfamily member 13b; tumor necrosis factor-like protein ZTNF4; UNQ401/PRO738; ZTNF4 | |
TNFSF13B | |
Primary | |
4°C, store in dark, DO NOT FREEZE! | |
TNFSF13B |
1D6 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Q9Y275 | |
10673 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.