Learn More
Invitrogen™ CD26 Monoclonal Antibody (202.36)

Description
202-36 Reacts with human CD26 (Dipeptidyl peptidase IV), a 110 kDa protein which occurs in the cell membrane of T-lymphocyte and is upregulated after activation. CD26 is also present on endothelial cells and also expressed on activated B-cells and natural killer cells and abundantly on epithelia. CD26 is implicated in a variety of biological functions including T-cell activation, cell adhesion with extracellular matrix such as fibronectin or collagens, and in HIV infection. Crosslinking of CD26 using this antibody dramatically enhances the anti-CD3-induced IL-2 production. In Western blotting, this mAb reacts with only glycosylated CD26, but not with deglycosylated froms. It does not prevent ADA binding to CD26. The mAb was clustered at the VIth WLDA.
Specifications
Specifications
| Antigen | CD26 |
| Applications | Flow Cytometry, Immunohistochemistry (Frozen), Western Blot |
| Classification | Monoclonal |
| Clone | 202.36 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | DPP4 |
| Gene Accession No. | P14740, P27487 |
| Gene Alias | ACT3; Activation molecule 3; ADABP; ADCP2; ADCP-2; ADCP-I; adenosine deaminase complexing protein; Adenosine deaminase complexing protein 2; bile canaliculus domain-specific membrane glycoprotein; CD26; Dipeptidyl peptidase 4; Dipeptidyl peptidase 4 60 kDa soluble form; Dipeptidyl peptidase 4 membrane form; Dipeptidyl peptidase 4 soluble form; Dipeptidyl peptidase IV; Dipeptidyl peptidase IV 60 kDa soluble form; Dipeptidyl peptidase IV membrane form; Dipeptidyl peptidase IV soluble form; dipeptidylpeptidase 4; dipeptidyl-peptidase 4; dipeptidyl-peptidase 4 (CD26, adenosine deaminase complexing protein 2); dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2); DPP IV; DPP4; Dpp-4; DPPIV; DPP-IV; GP110 glycoprotein; I79_016618; serine protease; T-cell activation antigen CD26; THAM; Thymocyte-activating molecule; TP103; WC10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.