Learn More
CD26 Mouse, anti-Human, R718, Clone: L272, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 752055
Description
The L272 monoclonal antibody specifically recognizes CD26 which is also known as Dipeptidyl peptidase IV (DPP IV), a serine protease. CD26 is a ∼120 kDa type II transmembrane glycoprotein that is encoded by DPP4 and belongs to the DPP4 activity and/or structure homologue (DASH) protein family. It is associated with the binding of the TAT transactivating protein of the HIV. CD26 and CD45 act in a costimulatory fashion on T lymphocytes. Present on peripheral blood T lymphocytes, the CD26 antigen is upregulated on phytohemagglutinin (PHA) and concanavalin A (Con A)-stimulated peripheral blood mononuclear cells (PBMCs). The CD26 antigen is found on approximately 50% of CD4 and approximately 30% of CD8 cells. It is also found on some mature thymocytes, B cells, natural killer (NK) cells, monocytes, macrophages, epithelial cells, EBV transformed B-cell lines, and hairy cell leukemia. Absolute numbers of CD4+CD26+ and CD8+CD26+ cells are reported to be lower in HIV-positive individuals. The CD26-CD4+ cell appears to be a reservoir for HIV. CD26 bright CD4 lymphocytes are CD25+ and CD45RO+ memory T lymphocytes.

Specifications
CD26 | |
Monoclonal | |
R718 | |
50 μg | |
Primary | |
Refrigerate Upon Arrival |
Flow Cytometry | |
L272 | |
Mouse | |
RUO | |
Human | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.