Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD261 (DR4) Monoclonal Antibody (DJR1), eBioscience™, Invitrogen™

Catalog No. 14664437
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-664-437 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-664-437 Supplier Invitrogen™ Supplier No. 14664437
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The DJR1 monoclonal antibody reacts with human DR4, also known as TRAIL-R1. DR4 binds to TRAIL, activates NF-kappaB, and induces TRAIL-mediated apoptosis. DR4 is expressed by a variety of human tumor cells and at very low level on human leukocytes. Expression has also been found inside the cell on some cell lines (Zhang et al). This has been confirmed with intracellular staining of Jurkat cell line. Please contact tech@ebioscience.com for more information. Applications Reported: This DJR1 antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: The DJR1 antibody has been tested by flow cytometric analysis of human DR4-transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD261 (DR4)
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone DJR1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene TNFRSF10A
Gene Accession No. O00220
Gene Alias APO2; CD261; cytotoxic TRAIL receptor; death receptor 4; DR4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF receptor superfamily member 10a; TNF-related apoptosis-inducing ligand receptor 1; TNFRSF10A; TRAIL R1; TRAIL receptor 1; TRAILR1; TRAIL-R1; TRAILR-1; tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a
Gene Symbols TNFRSF10A
Host Species Mouse
Purification Method Affinity chromatography
Quantity 2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8797
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.