Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD263 (TRAIL-R3) Mouse anti-Human, Alexa Fluor 647, Clone: B-D44, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565458
Description
CD263 (TRAIL-R3) Monoclonal antibody specifically detects CD263 (TRAIL-R3) in Human samples. It is validated for Flow cytometry.
Specifications
| CD263 (TRAIL-R3) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| 50 Tests | |
| Cell Biology | |
| Human | |
| Purified |
| Flow Cytometry | |
| B-D44 | |
| Vol. Per test: 5μL | |
| TRAIL-R3; TRAILR3; TRAIL Receptor 3; TNFRSF10C ; TR10C; DCR1; LIT; TRID | |
| Recombinant human TRAIL R3/Fc Protein | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
| IgG1 |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction