Learn More
Invitrogen™ CD263 (TRAIL-R3) Recombinant Rabbit Monoclonal Antibody (003), APC

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA546670
Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human TRAIL-R3 (CD263) (Met1-Pro235) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.
TRAIL-R3 (CD263) is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain and a transmembrane domain, but no cytoplasmic death domain. This receptor is not capable of inducing apoptosis, and is thought to function as an antagonistic receptor that protects cells from TRAIL-induced apoptosis. This gene was found to be a p53-regulated DNA damage-inducible gene. The expression of this gene was detected in many normal tissues but not in most cancer cell lines, which may explain the specific sensitivity of cancer cells to the apoptosis-inducing activity of TRAIL.
Specifications
CD263 (TRAIL-R3) | |
Recombinant Monoclonal | |
100 μg/mL | |
PBS with 0.5% BSA and 0.09% sodium azide | |
O14798 | |
TNFRSF10C | |
Recombinant protein ecoding Human CD263 (TRAIL-R3) (Met1-Pro235). | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry | |
003 | |
APC | |
TNFRSF10C | |
Antagonist decoy receptor for TRAIL/Apo-2L; CD263; cytotoxic TRAIL receptor-3; DCR1; DCR1-TNFR; Decoy receptor 1; decoy TRAIL receptor without death domain; LIT; lymphocyte inhibitor of TRAIL; MGC149501; MGC149502; TNF receptor superfamily member 10c; TNF-related apoptosis-inducing ligand receptor 3; TNFRSF10C; TRAIL receptor 3; TRAIL receptor without an intracellular domain; TRAILR3; TRAIL-R3; TRID; Tumor necrosis factor receptor superfamily member 10C; tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain; UNQ321/PRO366 | |
Rabbit | |
Protein A | |
RUO | |
8794 | |
4°C, store in dark, DO NOT FREEZE! | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.