Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CD264 (TRAIL-R4) Monoclonal Antibody (TRAIL-R4-01)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA119730
Description
This antibody reacts with an extracellular epitope of TRAIL-R4, a 42 kDa transmembrane protein expressed on various blood cells.
TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.
Specifications
CD264 (TRAIL-R4) | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q9UBN6 | |
TNFRSF10D | |
TRAIL-R4 (aa 1-210) - hIgGhc fusion protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
TRAIL-R4-01 | |
Unconjugated | |
TNFRSF10D | |
CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288 | |
Mouse | |
Protein A | |
RUO | |
8793 | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction