Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD264 (TRAIL-R4) Recombinant Rabbit Monoclonal Antibody (137)
GREENER_CHOICE

Catalog No. PIMA531248
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531248 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531248 Supplier Invitrogen™ Supplier No. MA531248
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human TRAIL R4/CD264/TNFRSF10D.

TRAIL (also known as Apo-2L) is a member of the tumor necrosis factor (TNF) ligand family that rapidly induces apoptosis in a variety of transformed cell lines. A fourth member of the emerging TRAIL receptor family, TRAIL-R4, has signaling capacity similar to that of TRAIL-R1 and TRAIL-R2 with respect to NF-kappaB activation, but differs in its inability to induce apoptosis. Like TRAIL-R1 and TRAIL-R2, this receptor shows widespread tissue expression. The human TRAIL-R4 gene has been mapped to chromosome 8p22-21, clustered with three other TRAIL receptors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD264 (TRAIL-R4)
Applications ELISA
Classification Recombinant Monoclonal
Clone 137
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene TNFRSF10D
Gene Accession No. Q9UBN6
Gene Alias CD264; DCR2; Decoy receptor 2; TNF receptor superfamily member 10d; TNF receptor-related receptor for TRAIL; TNF-related apoptosis-inducing ligand receptor 4; TNFRSF10D; TRAIL receptor 4; TRAIL receptor with a truncated death domain; TRAILR4; TRAIL-R4; TRUNDD; Tumor necrosis factor receptor superfamily member 10D; tumor necrosis factor receptor superfamily member 10d, decoy with truncated death domain; tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain; UNQ251/PRO288
Gene Symbols TNFRSF10D
Host Species Rabbit
Immunogen Recombinant Human TRAIL R4/CD264/TNFRSF10D protein (Met1-His211).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8793
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.