Learn More
Invitrogen™ CD265 (RANK) Monoclonal Antibody (R12-31), Biotin, eBioscience™
Rat Monoclonal Antibody
$285.00
Specifications
Antigen | CD265 (RANK) |
---|---|
Clone | R12-31 |
Concentration | 0.5 mg/mL |
Content And Storage | 4°C, do not freeze |
Applications | Flow Cytometry |
Description
Description: The R12-31 monoclonal antibody reacts with mouse RANK (Receptor activator of NF-kappaB), a recently cloned member of the TNFR superfamily with no significant homology to other members of this family. RANK ligand (RANKL/TRANCE/ OPGL) binds to RANK on dendritic cells, upregulates the expression of anti-apoptotic protein BcL-XL suggesting a role in dendritic cell survival. The cytoplasmic domain of RANK interacts with TRAF2, TRAF5 and TRAF6. Overexpression of RANK activates NF-kappaB and c-Jun-terminal kinase (JNK) pathways. Recent studies have shown that RANK interaction with TRAF6 activates NF-kappaB, whereas JNK activation is mediated through binding of RANK to TRAF2. Applications Reported: The R12-31 antibody has been reported for use in flow cytometric analysis. Applications Tested: This R12-31 antibody has been tested by flow cytometric analysis of mouse RANK-transfected cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
RANK is a member of the tumor necrosis factor receptor (TNFR) family. The full length mouse RANK cDNA encodes a type I transmembrane protein with an extracellular and a cytoplasmic domain. The extracellular domain contains two potential N-linked glycosylation sites. RANK shares significant amino acid homology with other members of the TNFR family in its extracellular four cysteine-rich repeats. Human and murine RANK share 81% amino acid identity in their extracellular domains.Specifications
CD265 (RANK) | |
0.5 mg/mL | |
Flow Cytometry | |
Biotin | |
Rat | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
O35305 | |
21934 | |
Primary | |
Affinity Chromatography |
R12-31 | |
4°C, do not freeze | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Mouse | |
TNFRSF11A | |
CD265; FEO; LOH18CR1; loss of heterozygosity, 18, chromosomal region 1; Ly109; mRANK; ODFR; OFE; OPTB7; Osteoclast differentiation factor receptor; OSTS; Paget disease of bone 2; PDB2; Rank; receptor activator of NF-kappaB; receptor activator of NF-KB; receptor activator of nuclear factor-kappa B; RGD1563614; TNF receptor superfamily member 11a; Tnfrsf11a; TRANCER; TRANCE-R; tumor necrosis factor receptor superfamily member 11A; tumor necrosis factor receptor superfamily member 11a, NFKB activator; tumor necrosis factor receptor superfamily, member 11a, NFKB activator | |
TNFRSF11A | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.