Learn More
CD266 Mouse anti-Human, Brilliant Violet 786, Clone: ITEM-1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 743390
Description
The ITEM-1 monoclonal antibody specifically binds to CD266, which is also known as TWEAK Receptor (TWEAK-R/TWEAKR), or Fibroblast growth factor inducible 14 (FGF-inducible 14, FN14). CD266 is a ñ14 kDa type I transmembrane protein that is the 12A family member of the tumor necrosis factor receptor superfamily (TNFRSF12A). It is expressed at low levels in a variety of tissues including, heart, placenta, lung, muscle, and pancreas. CD266 is relatively highly expressed on HUVEC cells and certain tumor cell lines. TWEAK-induced CD266-mediated signaling can play roles in inflammation, the induction of apoptosis in certain cell types, the proliferation and migration of endothelial cells, and can promote angiogenesis within healthy and diseased tissues, eg, tumors. The ITEM-1 antibody can reportedly induce the proliferation of HUVEC cells and the death of certain TWEAK-sensitive tumor target cell lines. CD266 binds TWEAK (CD255) and can signal intracellularly by interactions with TRAF1, 2, and 3 cytoplasmic proteins leading to NF-κB activation.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
| CD266 | |
| ITEM-1 | |
| Brilliant Violet 786 | |
| TWEAKR, TWEAK-R, TWEAK Receptor, TNFRSF12A, FGF-inducible 14, FN14 | |
| Human | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.