Learn More
Invitrogen™ CD266 (TWEAK Receptor) Monoclonal Antibody (ITEM-1), eBioscience™, Invitrogen™

Description
The ITEM-1 mAb reacts with human TWEAK Receptor/Fn14 (fibroblast growth factor-inducible 14 kDa protein). Fn14 is distantly related to the TNFR family, containing one cysteine-rich domain in the extracellular region and a TNFR-associated factor binding domain but does not contain a death domain (DD) cytoplasmic region. Fn14 plays a role in TWEAK-induced endothelial cell migration, proliferation, and angiogenesis. TWEAK-induced cell death via Fn14 includes both apoptosis and necrosis and can be blocked by an anti-TWEAK antibody, CARL-1. Fn14 is expressed on HUVEC and in some cancer tissues but not on freshly isolated PBMCs. Fn14 mRNA expression has been identified during liver regeneration.
Specifications
Specifications
| Antigen | CD266 (TWEAK Receptor) |
| Applications | Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | ITEM-1 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | TNFRSF12A |
| Gene Accession No. | Q9NP84 |
| Gene Alias | AI255180; C87282; CD266; FGF-inducible 14; Fgfrp2; fibroblast growth factor regulated protein 2; fibroblast growth factor-inducible immediate-early response protein 14; Fibroblast growth factor-regulated protein 2; FN14; HPIP; TNF receptor superfamily member 12A; TNFRSF12A; Tumor necrosis factor receptor superfamily member 12A; tumor necrosis factor receptor superfamily, member 12A; TweakR; TWEAK-R; Tweak-receptor; type I transmembrane protein Fn14 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.