Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD267 (TACI) Monoclonal Antibody (11H3), PE, eBioscience™
GREENER_CHOICE

Catalog No. 5011257
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-57 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-112-57 Supplier Invitrogen™ Supplier No. 12921742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Description: The 11H3 antibody reacts with human TACI, also known as tumor necrosis factor receptor 13B. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kB and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases. TACI mRNA and protein are found predominantly in B-cells and correlate with BLyS binding in a panel of B-cell lines. TACI interacts with nanomolar affinity with the BLyS-related tumor necrosis factor homologue APRIL for which no clear in vivo role has been described. 11H3 does not block binding of BAFF/BLyS to TACI. Applications Reported: This 11H3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 11H3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Members in the TNF superfamily regulate immune responses and induce apoptosis. Two novel members in the TNF family were recently identified and designated BAFF/BLyS/TALL-1/THANK/zTNF4 and April/TALL-2, respectively. BAFF was characterized as a B cell activator since it induced B cell proliferation and immunoglobulin secretion. April regulates immunological and non-immunological cell growth. Three receptors, BCMA, TACI, and BAFF-R, for BAFF and April were recently identified. TACI, like BCMA, binds BAFF and April. TACI and its ligands regulate humoral immune responses, activate NF-kappa-B and c-jun N-terminal kinase, and are involved in B cell associated autoimmune diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD267 (TACI)
Applications Flow Cytometry
Classification Monoclonal
Clone 11H3
Concentration 5 μL/Test
Conjugate PE
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene TNFRSF13B
Gene Accession No. O14836
Gene Alias 1200009E08Rik; CD267; CVID; CVID2; IGAD2; NEWGENE_1595998; RYZN; TACI; TNF receptor superfamily member 13B; Tnfrsf13b; TNFRSF14B; transmembrane activator and calcium-modulating and cyclophilin ligand interactor; transmembrane activator and CAML interactor; tumor necrosis factor receptor 13B; tumor necrosis factor receptor superfamily member 13B; tumor necrosis factor receptor superfamily, member 13B
Gene Symbols TNFRSF13B
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23495
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.