Learn More
CD268 (BAFF-R) Rat anti-Mouse, Brilliant Violet 510, Clone: 7H22-E16, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 742866
Description
The 7H22-E16 monoclonal antibody specifically binds to CD268, which is also known as, B cell-activating factor receptor (BAFF-R), or BAFF Receptor 3 (BR3). CD268 is a type III transmembrane protein that is likewise known as Tumor necrosis factor receptor superfamily member 13C (Tnfrsf13c). CD268/BAFF-R is a receptor for CD257 (also known as, BAFF, Blys, TALL-1, or THANK) and is expressed on B cells and a subset of activated/memory CD4+ T cells. B cells express two other BAFF receptors, CD267/TACI, or CD269/BCMA, at various times during their differentiation. CD268/BAFF-R expression starts at the transitional stage of B cell development and increases throughout B cell maturation. CD267/BAFF-R mediates most BAFF-dependent functions including B cell survival, the activation of B cell proliferation and immunoglobulin secretion, and the costimulation of T cells. Overexpression of BAFF in mice and humans is associated with autoimmunity, whereas CD268/BAFF-R deficiency may cause severe impairment in humoral responses.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405nm and Em Max at 510nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser. BV510 is a polymer-based dye that is brighter than BD Horizon V500. Due to similar excitation and emission properties, BD Horizon BV510 and BD Horizon V500 cannot be used simultaneously.
Specifications
CD268 (BAFF-R) | |
7H22-E16 | |
Brilliant Violet 510 | |
Tnfrsf13c, TR13C, BAFF-R, Baffr, Bcmd, Bcmd-1, BLyS receptor 3, Br3, Lvis22 | |
Mouse BAFF-R Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.