Learn More
CD268 (BAFF-R) Rat anti-Mouse, Brilliant Violet 605, Clone: 7H22-E16, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 742867
Description
The 7H22-E16 monoclonal antibody specifically binds to CD268, which is also known as, B cell-activating factor receptor (BAFF-R), or BAFF Receptor 3 (BR3). CD268 is a type III transmembrane protein that is likewise known as Tumor necrosis factor receptor superfamily member 13C (Tnfrsf13c). CD268/BAFF-R is a receptor for CD257 (also known as, BAFF, Blys, TALL-1, or THANK) and is expressed on B cells and a subset of activated/memory CD4+ T cells. B cells express two other BAFF receptors, CD267/TACI, or CD269/BCMA, at various times during their differentiation. CD268/BAFF-R expression starts at the transitional stage of B cell development and increases throughout B cell maturation. CD267/BAFF-R mediates most BAFF-dependent functions including B cell survival, the activation of B cell proliferation and immunoglobulin secretion, and the costimulation of T cells. Overexpression of BAFF in mice and humans is associated with autoimmunity, whereas CD268/BAFF-R deficiency may cause severe impairment in humoral responses.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
CD268 (BAFF-R) | |
7H22-E16 | |
Brilliant Violet 605 | |
Tnfrsf13c, TR13C, BAFF-R, Baffr, Bcmd, Bcmd-1, BLyS receptor 3, Br3, Lvis22 | |
Mouse BAFF-R Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.