Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD268 (BAFF-R) Rat anti-Mouse, Brilliant Violet 650, Clone: 7H22-E16, BD Optibuild™

Catalog No. BDB742868 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB742868 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB742868 Supplier BD Biosciences Supplier No. 742868
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 7H22-E16 monoclonal antibody specifically binds to CD268, which is also known as, B cell-activating factor receptor (BAFF-R), or BAFF Receptor 3 (BR3). CD268 is a type III transmembrane protein that is likewise known as Tumor necrosis factor receptor superfamily member 13C (Tnfrsf13c). CD268/BAFF-R is a receptor for CD257 (also known as, BAFF, Blys, TALL-1, or THANK) and is expressed on B cells and a subset of activated/memory CD4+ T cells. B cells express two other BAFF receptors, CD267/TACI, or CD269/BCMA, at various times during their differentiation. CD268/BAFF-R expression starts at the transitional stage of B cell development and increases throughout B cell maturation. CD267/BAFF-R mediates most BAFF-dependent functions including B cell survival, the activation of B cell proliferation and immunoglobulin secretion, and the costimulation of T cells. Overexpression of BAFF in mice and humans is associated with autoimmunity, whereas CD268/BAFF-R deficiency may cause severe impairment in humoral responses.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD268 (BAFF-R)
Classification Monoclonal
Clone 7H22-E16
Concentration 0.2mg/mL
Conjugate Brilliant Violet 650
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Tnfrsf13c, TR13C, BAFF-R, Baffr, Bcmd, Bcmd-1, BLyS receptor 3, Br3, Lvis22
Host Species Rat
Immunogen Mouse BAFF-R Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.