Learn More
Invitrogen™ CD268 (BAFF Receptor) Monoclonal Antibody (8A7), PE, eBioscience™

Description
Description: The 8A7 monoclonal antibody reacts with the human BAFF receptor, also known as B lymphocyte stimulator (BLyS protein) receptor, which is expressed on B cells. The ligand for this receptor, BAFF (B cell-activating factor of the TNF family) is a B cell survival factor and regulates CD21/35 and CD23 expression. Interaction of this ligand with its receptor causes elevated CD21/35 and CD23 expression, whereas receptor blockade has been shown to down-modulate expression. Applications Reported: The 8A7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 8A7 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.00375 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: μ8 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | CD268 (BAFF Receptor) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 8A7 |
| Concentration | 5 μL/Test |
| Conjugate | PE |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | TNFRSF13C |
| Gene Accession No. | Q96RJ3 |
| Gene Alias | 2010006P15Rik; B cell-activating factor receptor; BAFF receptor; BAFFR; BAFF-R; B-cell maturation defect; B-cell maturation defect 1; B-cell-activating factor receptor; Bcmd; Bcmd1; Bcmd-1; BLyS receptor 3; BR3; BROMIX; CD268; CVID4; Lvis22; prolixin; RGD1560810; TNF receptor superfamily member 13C; Tnfrsf13c; Tumor necrosis factor receptor superfamily member 13C; tumor necrosis factor receptor superfamily, member 13C |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.