Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD268 (BAFF Receptor) Mouse anti-Human, Alexa Fluor 647, Clone: 11C1, BD Pharmingen™
Mouse Monoclonal Antibody
$329.50
Specifications
| Antigen | CD268 (BAFF Receptor) |
|---|---|
| Clone | 11C1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Alexa Fluor 647 |
Description
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow cytometry.
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow cytometry.Specifications
| CD268 (BAFF Receptor) | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human BAFF-R Transfected Cell Line | |
| Primary | |
| Vol. Per test: 5μL |
| 11C1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BAFF Receptor; BAFF-R; BLyS receptor 3; BR3; TNFRSF13C | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title