Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD268 (BAFF Receptor) Mouse anti-Human, Brilliant Violet 786, Clone: 11C1, BD Optibuild™

Catalog No. BDB743574 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743574 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743574 Supplier BD Biosciences Supplier No. 743574
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 11C1 monoclonal antibody recognizes CD268 which is also known as the B cell activating factor receptor (BAFF-R). CD268 is a type III transmembrane protein of approximately 184 residues. BAFF-R is one of the receptors for B-cell activating factor (BAFF), a member of the TNF family of proteins. BAFF is a key regulator for B cell differentiation and critical in regulating survival and activation of peripheral B cell populations. Experiments in the mouse model show that interaction of BAFF with BAFF-R promotes NF-κB activity. Overexpression of BAFF results in an expanded B cell compartment and autoimmunity in mice. Mice injected with BAFF-neutralizing-Fc (BAFF-R-Fc) protein showed reduced NF-κB activation, blocking BAFF-induced B cell proliferation. Reports suggest that the BAFF-BAFF-R interaction may lead to a better understanding of autoimmune disorders.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD268 (BAFF Receptor)
Classification Monoclonal
Clone 11C1
Concentration 0.2mg/mL
Conjugate Brilliant Violet 786
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias BAFF Receptor, BAFF-R, BLyS receptor 3, BR3, TNFRSF13C
Host Species Mouse
Immunogen Human BAFF-R Transfected Cell Line
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.