Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD27 Ab-2, Mouse Monoclonal Antibody

Catalog No. MS1468S1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
500 μL
7 mL
Form:
Concentrated
Prediluted
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1468S1 500 μL Concentrated
MS1468S0 100 μL Concentrated
MS1468R7 7 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MS1468S1 Supplier Epredia™ Supplier No. MS1468S1

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD27 Ab-2, Mouse Monoclonal Antibody.

CD27 (also known as S152 or T14) is expressed on the majority of peripheral T cells, medullary thymocytes, memory-type B cells, and natural killer cells. It is a transmembrane phosphoglycoprotein that belongs to the tumor necrosis factor receptor (TNFR) superfamily. CD27 binds to its ligand CD70, a member of the TNF family, and induces T-cell costimulation and B-cell activation. It also interacts with TRAFs and is involved in activation of NFδB and SAPK/JNK and induces apoptosis.

Host Species: Mouse

Clone: 137B4

Isotype: IgG1

Species Reactivity: Human. Others not tested.

Immunogen: Recombinant human CD27 protein

Molecular Weight: 120 kDa

Positive Control: Tonsil

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen CD27 Ab-2
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 137B4
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human CD27 protein
Purification Method Tonsil
Quantity 500 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.