Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (301731), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2738X
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow CytometrySpecifications
| CD27 Ligand/TNFSF7/CD70 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
| Mouse myeloma cell line NS0-derived recombinant human CD27 Ligand/TNFSF7, Gln45-Pro193, Accession # AAA36175 | |
| 0.1 mL | |
| Apoptosis | |
| 970 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| 301731 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction