Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TAN 1-7), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250150V
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for Flow Cytometry, ELISA.Specifications
CD27 Ligand/TNFSF7/CD70 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD70 | |
Recombinant mouse CD27 Ligand/TNFSF7/CD70 (extracellular domain, aa residues 41-195) | |
0.1 mL | |
Apoptosis | |
970 | |
Mouse | |
Purified |
Flow Cytometry, ELISA | |
TAN 1-7 | |
Flow Cytometry, ELISA | |
CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
This antibody is a non-depleting mAb that blocks the CD70-CD27 interaction. | |
Store at 4C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction