Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27 Ligand/TNFSF7/CD70 Antibody (TNFS7/1026), PerCP , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311587PCP
Description
CD27 Ligand/TNFSF7/CD70 Monoclonal antibody specifically detects CD27 Ligand/TNFSF7/CD70 in Human samples. It is validated for Flow CytometrySpecifications
CD27 Ligand/TNFSF7/CD70 | |
Monoclonal | |
PerCP | |
CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
Recombinant human CD27 Ligand/TNFSF7/CD70 protein (Uniprot: P32970) | |
0.1 mL | |
Apoptosis | |
970 | |
Store at 4°C in the dark. | |
IgG1 κ |
Flow Cytometry | |
TNFS7/1026 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction