Learn More
CD27 Monoclonal Antibody (DEN-3), PE, Exalpha

Mouse Monoclonal Antibody
Supplier: Vector Laboratories 0274100TESTS


Description
Flow Cytometry PBMC: Add 10 μL of MAB/10^6 PBMC in 100 μL PBS. Mix gently and incubate for 15 minutes at 2 to 8C. Wash twice with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. Flow Cytometry Whole Blood: Add10 μL of MAB/100 μL of whole blood. Mix gently and incubate for 15 minutes at room temperature 20C. Lyse the whole blood. Wash once with PBS and analyze or fix with 0.5% v/v of paraformaldehyde in PBS and analyze. See instrument manufacturer instructions for Lysed Whole Blood and Immunofluorescence analysis with a flow cytometer or microscope.
CD27 Monoclonal antibody specifically detects CD27 in Human samples. It is validated for Flow CytometrySpecifications
CD27 | |
Monoclonal | |
PE | |
CD27 | |
CD antigen 27; CD27; CD27 antigen; CD27 molecule; CD27L receptor; LPFS2; S152; S152. LPFS2; sCD27; soluble CD27; T cell activation antigen S152; T14; T-cell activation antigen CD27; TNFRSF7; TNFSF7; Tp55; Tumor necrosis factor receptor superfamily member 7; tumor necrosis factor receptor superfamily, member 7 | |
Protein A/G | |
RUO | |
939 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry | |
DEN-3 | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.2 | |
P26842 | |
Mouse | |
100Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.