Learn More
Description
Specifications
Specifications
| Antigen | CD27 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | M-T271 |
| Conjugate | Brilliant Violet 650 |
| Description | Vol. Per test: 5μL |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | TNFRSF7; TNF receptor superfamily, member 7; T14; Tp55; S152 |
| Host Species | Mouse |
| Immunogen | Human T-CLL cells |
| Show More |
For Reseach Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.