Learn More
CD27 Mouse anti-Human, PE, Clone: M-T271, BD
Mouse Monoclonal Antibody
$88.40 - $243.00
Specifications
Antigen | CD27 |
---|---|
Clone | M-T271 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Description
The M-T271 monoclonal antibody specifically binds to CD27. CD27 presents asa type I transmembrane, disulphide-linked 110 kDa homodimer comprised of two polypeptide chains. The CD27 molecule is a lymphocyte-specific member of the TNF/NGF-R family, and is expressed on a subset of human thymocytes and on the majority of mature T lymphocytes, activated B cells and NK cells. CD27 is highly induced on T cells after TCR stimulation. CD27 binds to CD70 (also known as, CD27 ligand or CD27L) and may be involved in cellular interaction of T and B lymphocytes.
Specifications
CD27 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified |
M-T271 | |
Monoclonal | |
Mouse | |
Human | |
Human T-CLL cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.