Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (133), Alexa Fluor™ 405

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290424AF405
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for ELISA.Specifications
CD27/TNFRSF7 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human CD27 Human TNFRSF8 Human CD40 Human DR6 Human TNFRSF10D Mouse HVEM Mouse FAS Mouse TNFRSF10b Human 4-1BB Human TNFRI Human TNFRII Human RELT Human FAS | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
133 | |
ELISA | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD27/TNFRSF7 (NP_001028298.1; Met 1-Arg 182). | |
0.1 mL | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity | |
939 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction