Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (150), DyLight 680

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289228FR
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Western Blot, ELISA.Specifications
CD27/TNFRSF7 | |
Monoclonal | |
DyLight 680 | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD27/TNFRSF7 (P26842; Met1-Ile192). | |
0.1 mL | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity | |
939 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
150 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction