Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (LG.7F9), FITC, Novus Biologicals™

Armenian Hamster Monoclonal Antibody
$340.50 - $780.00
Specifications
Antigen | CD27/TNFRSF7 |
---|---|
Clone | LG.7F9 |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14402105
![]() |
Novus Biologicals
NBP1440210.05MG |
0.05 mg |
Each for $340.50
|
|
|||||
NBP14402101
![]() |
Novus Biologicals
NBP1440210.1MG |
0.1 mg |
Each for $622.00
|
|
|||||
NBP1440215
![]() |
Novus Biologicals
NBP1440210.5MG |
0.5 mg |
Each for $780.00
|
|
|||||
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
CD27/TNFRSF7 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Armenian Hamster | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
939 | |
The immunogen for this antibody was CD27. | |
Primary | |
Store at 4C. Do not freeze. |
LG.7F9 | |
Flow Cytometry | |
FITC | |
RUO | |
Human, Mouse, Rat | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
CD27 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title