Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (O323), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP143426AF350
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry.Specifications
CD27/TNFRSF7 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
CD27 | |
The immunogen for this antibody was CD27. | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
O323 | |
Flow Cytometry | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
0.1 mL | |
939 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction