Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD27/TNFRSF7 Antibody (O323), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$206.50 - $499.50

Specifications

Antigen CD27/TNFRSF7
Clone O323
Dilution Flow Cytometry 1:10-1:1000
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14342603
SDP
View Documents
Novus Biologicals
NBP1434260.025MG
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP14342601
SDP
View Documents
Novus Biologicals
NBP1434260.1MG
0.1 mg
Each for $499.50
Only null left
Add to Cart
 
Description

Description

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD27/TNFRSF7
Flow Cytometry 1:10-1:1000
Monoclonal
Mouse
Apoptosis, Cancer, Immunology, Innate Immunity
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
939
The immunogen for this antibody was CD27.
Primary
Store at 4C. Do not freeze.
O323
Flow Cytometry
Unconjugated
RUO
Human
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
CD27
IgG1 κ
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.