Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (PG124A), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260874
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
CD27/TNFRSF7 | |
Monoclonal | |
1 mg/mL | |
Flow Cytometry 1:10 - 1:1000 | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
Mouse | |
Unpurified | |
Apoptosis, B Cell Development and Differentiation Markers, Cancer, Immunology, Innate Immunity | |
939 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Flow Cytometry, Immunohistochemistry, Functional Assay | |
PG124A | |
Unconjugated | |
Ascites with 0.09% Sodium Azide | |
CD27 | |
Pig mononuclear cells | |
0.1 mg | |
Primary | |
Pig | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction