Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD27/TNFRSF7 Antibody (RM27-3E5) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP275199
Description
CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Neutralization.Specifications
CD27/TNFRSF7 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
CD27 | |
This antibody was raised by immunising mouse with mouse CD27-human IgG1 Fc fusion protein. Isolated popliteal lymph node cells were then fused with P3U1 myeloma cells to produce stable hybridomas. | |
0.2 mg | |
939.0 | |
Mouse | |
Purified |
Flow Cytometry, Immunoprecipitation, Neutralization | |
RM27-3E5 | |
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, Neutralization | |
CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7 | |
Rat | |
Protein A purified | |
Primary | |
This antibody is specific for the extracellular domain of murine CD27, a TNF-receptor superfamily member. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction