Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD272 (BTLA) Mouse, anti-Human, R718, Clone: J168-540, BD OptiBuild™

Catalog No. BDB752231 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752231 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752231 Supplier BD Biosciences Supplier No. 752231
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The J168-540 monoclonal antibody specifically binds to human CD272, also known as BTLA (B and T lymphocyte attenuator), an inhibitory receptor expressed in bone marrow and thymus on developing B and T cells. In the periphery, BTLA is expressed by B cells, T cells, bone marrow-derived dendritic cells, and macrophages. After T cell activation, BTLA appears to be expressed at higher levels on Th1 cell populations than in Th2 cell populations. Upon binding the herpesvirus entry mediator (HVEM/LIGHT-R/CD270), CD272 undergoes tyrosine phosphorylation and inhibits T cell proliferation in a BTLA-dependent manner. CD272 has structural similarities to two other lymphocyte inhibitory receptors, CTLA-4 and PD-1 and is a member of the CD28-like family of coreceptors. Based on these observations, CD272 is considered to be a negative regulator of lymphocyte activation and/or function.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD272 (BTLA)
Applications Flow Cytometry
Classification Monoclonal
Clone J168-540
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.