Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD272 (BTLA) Mouse anti-Mouse, BV421, Clone: 6F7/BTLA (also known as 6F7), BD Horizon™
Mouse Monoclonal Antibody
$278.50
Specifications
| Antigen | CD272 (BTLA) |
|---|---|
| Clone | 6F7/BTLA (also known as 6F7) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD272 (BTLA) Monoclonal antibody specifically detects CD272 (BTLA) in Mouse samples. It is validated for Flow cytometrySpecifications
| CD272 (BTLA) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 208154 | |
| IgG1 κ | |
| Affinity Chromatography |
| 6F7/BTLA (also known as 6F7) | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Mouse | |
| B- and T-lymphocyte attenuator; Btla | |
| C57BL/6 BTLA Ig Domain Protein | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title