Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD272 (BTLA) Mouse anti-Mouse, PE, Clone: 6F7/BTLA (also known as 6F7), BD Pharmingen™
Mouse Monoclonal Antibody
$173.40 - $372.50
Specifications
| Antigen | CD272 (BTLA) |
|---|---|
| Clone | 6F7/BTLA (also known as 6F7) |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Description
CD272 (BTLA) Monoclonal antibody specifically detects CD272 (BTLA) in Mouse samples. It is validated for Flow cytometry (Routinely Tested)Specifications
| CD272 (BTLA) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| C57BL/6 BTLA Ig Domain Protein | |
| Primary | |
| The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. |
| 6F7/BTLA (also known as 6F7) | |
| Flow Cytometry | |
| PE | |
| Mouse | |
| Mouse | |
| B- and T-lymphocyte attenuator; Btla | |
| IgG1 κ | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title