Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD273 Mouse anti-Human, RB780, Clone: MIH18, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 755831
This item is not returnable.
View return policy
Description
CD273 Monoclonal antibody specifically detects CD273 in Human samples. It is validated for Flow Cytometry
Specifications
CD273 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
50 μg | |
Primary | |
Human | |
IgG1 κ |
Flow Cytometry | |
MIH18 | |
RB780 | |
PD-1 ligand 2; PD-L2; B7-DC; PDCD1LG2; PDCD1 ligand 2; PDCD1L2; PDL2; Btdc | |
Human CD273 Transfected Cell Line | |
RUO | |
80380 | |
Store undiluted at 4°C and protected from prolonged exposure to light.Do not freeze.The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction