Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD273 (PD-L2), Rat Anti-Mouse, RB613, Clone: MIH37, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 758326
Description
CD273 (PD-L2) Monoclonal antibody specifically detects CD273 (PD-L2) in Mouse (Tested in Development) samples. It is validated for Flow Cytometry
Specifications
CD273 (PD-L2) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
50 μg; | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
MIH37 | |
RB613 | |
B7-DC; Btdc; PD-1 ligand 2; PD-L2; PDCD1 ligand 2; programmed cell death 1 ligand 2 | |
Mouse B7-DC-transfected Cell Line | |
RUO | |
58205 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction