Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274 Mouse anti-Human, PE, Clone: MIH1, BD

Catalog No. BDB557924 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557924 100 Tests
BDB561787 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB557924 Supplier BD Biosciences Supplier No. 557924
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PD-L1 and PD-L2 are newly discovered members of the B7 family are the ligands for the program death 1 (PD-1) receptor. They are expressed on immature dendritic cells and mature dendritic cells. PD-L1, also called B7-H1, is expressed on antigen-presenting cells, including IFN- γ-stimulated monocytes, and activated human and murine dendritic cells. Monoclonal antibodies that block PD-L1 and PD-L2 on dendritic cells result in enhanced T cell proliferation and cytokine production. PD-L1 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. Studies show overlapping functions of PD-L1 and PD-L2 and indicate an important role for the PD-L:PD-1 pathway in regulating T cell responses.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274
Applications Flow Cytometry
Classification Monoclonal
Clone MIH1
Conjugate PE
Description B7-H1, PD-L1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.