Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274, NA/LE Mouse anti-Human, Unlabeled, Clone: MIH1, BD

Catalog No. BDB565959 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB565959 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB565959 Supplier BD Biosciences Supplier No. 565959
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MIH1 monoclonal antibody specifically binds to CD274, which is also known as, B7 homolog 1 (B7-H1), Programmed cell death 1 ligand 1 (PDCD1 ligand, PDCD1L1, PDCD1LG1), or Programmed death ligand 1 (PD-L1, PDL1). CD274 and PD-L2 (CD273) are type I transmembrane glycoproteins that belong to the B7 family and serve as ligands for CD279 (Program Death 1/PD-1). CD274 is expressed on antigen-presenting cells including activated monocytes/macrophages and dendritic cells, as well as, activated T cells, and keratinocytes. CD274 is also expressed on placental trophoblasts, myocardial endothelium, cortical thymic epithelial cells, and on most carcinomas. CD274 plays an important role in regulating T cell responses. The MIH1 antibody blocks CD279 binding to CD274 and can enhance the proliferation and cytokine production of activated T cells.

NA/LE
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274, NA/LE
Classification Monoclonal
Clone MIH1
Concentration 1.0mg/mL
Conjugate Unconjugated
Formulation No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay.
Gene Alias B7-H1, B7-H, PD-L1, PDL1, PDCD1 ligand 1, PDCD1L1, PDCD1LG1
Host Species Mouse
Immunogen Human CD274 Transfected Cell Line
Purification Method Affinity Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.