Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cd274 Rabbit anti-Human, Clone:RM320, Abnova™

Catalog No. 89333282 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-282 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-282 Supplier Abnova Corporation Supplier No. MAB16935
Add to Cart
Add to Cart

Rabbit monoclonal antibody raised against huamn Cd274.

Specifications

Antigen Cd274
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone RM320
Conjugate Unconjugated
Description CD274 antigen
Dilution Immunocytochemistry (1:200-500)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1000)
Western Blot (1:500-1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Gene Cd274
Gene Alias B7-H1, PD-L1, Pdcd1l1, Pdcd1lg1
Host Species Rabbit
Immunogen A peptide corresponding to C-terminus of human Cd274.
Quantity 100 μL
Gene ID (Entrez) 60533
Test Specificity This antibody reacts to human PD-L1 (B7-H1).
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.