Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD274 Rat anti-Mouse, Clone: MIH5, BD Pharmingen™

Catalog No. BDB567839 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB567839 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB567839 Supplier BD Biosciences Supplier No. 567839
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD274 Monoclonal antibody specifically detects CD274 in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD274
Applications Flow Cytometry
Classification Monoclonal
Clone MIH5
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias B7-H1, PD-L1; PD1L1; Programmed death ligand 1
Host Species Rat
Immunogen DBA/2 mouse lymphoma L5178Y transfected with Pdcd1lg1 cDNA
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Form Liquid
Isotype IgG2a λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.