Learn More
Description
Specifications
Specifications
| Antigen | CD274 |
| Applications | Single Cell 3' Sequencing |
| Classification | Monoclonal |
| Clone | Clone MIH5 |
| Conjugate | Oligo |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Gene Alias | B7-H1, PD-L1; PD1L1; Programmed death ligand 1 |
| Host Species | Rat |
| Immunogen | DBA/2 mouse lymphoma L5178Y transfected with Pdcd1lg1 cDNA |
| Quantity | 25 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.