Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD275 (B7-H2) Monoclonal Antibody (4H2B1), Invitrogen™

Mouse Monoclonal Antibody
$285.50 - $493.00
Specifications
| Antigen | CD275 (B7-H2) |
|---|---|
| Clone | 4H2B1 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
PIMA543376
|
Invitrogen™
MA543376 |
50 μL |
Each for $285.50
|
|
|||||
PIMA543024
![]() |
Invitrogen™
MA543024 |
100 μL |
Each for $493.00
|
|
|||||
Description
Specificity: Human ICOS Ligand/B7-H2/ICOSLG
Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.Specifications
| CD275 (B7-H2) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand | |
| ICOSLG | |
| IgG1 | |
| Protein A | |
| Antibody |
| 4H2B1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| O75144 | |
| 23308 | |
| Recombinant Human ICOS Ligand/B7-H2/ICOSLG protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| ICOSLG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title