Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD275 (B7-H2) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596572
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596572 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596572 Supplier Invitrogen™ Supplier No. PA596572
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DTQEKEVRAM VGSDVELSCA CPEGSRFDLN DVYVYWQTSE SKTVVTYHIP QNSSLENVDS RYRNRALMSP AGMLRGDFSL RLFNVTPQDE QKFHCLVLSQ SLGFQEVLSV EVTLHVAANF SVPVVSAPHS PSQDELTFTC TSINGYPRPN VYWINKTDNS LLDQALQNDT VFLNMRGLYD VVSVLRIART PSVNIGCCIE NVLLQQNLTV GSQTGNDIGE RDKITENPVS TGEKNAAT; Positive Samples: HL-60, BT-474, Mouse heart, Mouse brain, Rat spleen; Cellular Location: Membrane, Single-pass type I membrane protein.

Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD275 (B7-H2)
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.94 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene ICOSLG
Gene Accession No. O75144, Q9JHJ8
Gene Alias AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand
Gene Symbols Icosl, ICOSLG
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 19-256 of human ICOSL (NP_0560741).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23308, 499415, 50723
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.