Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD275 Mouse, anti-Human, R718, Clone: 2D3/B7-H2, BD OptiBuild™

Catalog No. BDB751883 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751883 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751883 Supplier BD Biosciences Supplier No. 751883
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 2D3 monoclonal antibody specifically binds to CD275, which is also known as B7 homolog 2 (B7-H2/B7H2) and B7-related protein 1 (B7RP-1). CD275 is a type I transmembrane glycoprotein that belongs to the Ig superfamily. CD275 is likewise known as Inducible T-cell co-stimulator ligand (ICOS Ligand /ICOSLG/ICOS-L). It can bind to CD278 (ICOS) and costimulate the proliferation and cytokine production of antigen-stimulated human T cells. CD275 is variable expressed by subsets of B cells, monocytes, macrophages, dendritic cells and endothelial cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD275
Applications Flow Cytometry
Classification Monoclonal
Clone 2D3/B7-H2
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.