Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CD276 (B7-H3) Monoclonal Antibody (M3.2D7), PE, eBioscience™, Invitrogen™
GREENER_CHOICE

Catalog No. 5011042
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-110-42 100 μg
50-110-41 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-110-42 Supplier Invitrogen™ Supplier No. 12597382
Add to Cart
Edge
Add to Cart

Rat Monoclonal Antibody

Description: The M3.2D7 monoclonal antibody was generated against and reacts with mouse B7-H3, a member of the B7 family of the Ig superfamily proteins. B7-H3 is reported to be expressed by antigen presenting cells and developing bone during embryogenesis. The ligand for B7-H3 has not yet been identified and it is thought that B7-H3 down-regulates T cell functions by engaging an unknown counter receptor on T cells. M3.2D7 stains mouse B7-H3 transfected cells and not mouse spleen cells. M3.2D7 also stains human B7-H3 transfected cells. Applications Reported: The M3.2D7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This M3.2D7 antibody is offered in 2 formats: - μg size: has been tested by flow cytometric analysis of mouse B7-H3 transfected cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by flow cytometric analysis of mouse B7-H3 transfected cells. This can be used at 5 μL (0.25 μg) per test.

CD276 is a member of the B7 family of co-stimulatory molecules also known as B7-H3. CD276 is a type I transmembrane protein that induces the proliferation of CD4+ and CD8+ T cells, enhances the generation of cytotoxic T cells and selectively stimulates the production of interferon gamma. Expression of CD276 can be induced on dendritic cells and monocytes by inflammatory cytokines, and is also widely expressed in peripheral tissues including the heart, kidney, testis and colon. In humans, CD276 exists as two isoforms which result from gene duplication and differential splicing. CD276 is reported to have therapeutic potential for the regulation of cell-mediated immune responses to cancer, particularly in conjunction with anti-angiogenic therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD276 (B7-H3)
Applications Flow Cytometry
Classification Monoclonal
Clone M3.2D7
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene CD276
Gene Accession No. Q8VE98
Gene Alias 4Ig-B7-H3; 6030411F23Rik; AU016588; B7 homolog 3; B7h3; B7-H3; B7RP-2; CD276; CD276 antigen; CD276 molecule; CD276 molecule tv1; costimulatory molecule; PSEC0249; sB7 H3; sB7H3; sCD276; soluble B7 H3; soluble B7H3; soluble CD276; UNQ309/PRO352
Gene Symbols CD276
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102657
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.